Learn More
GSC Go Science Crazy Optical Bench
SCI_ED
Catalog No. p-6765099 Shop All GSC Products
Encompass
Change view
Click to view available options
Length (Metric) Focal:
100 cm
Includes:
With meter stick
Without meter stick
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Focal Includes
S13648 100 cm With meter stick
S13649 - Without meter stick
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Not a toy. Product intended for ages 13 and up.
Catalog No. S13648 Supplier GSC Go Science Crazy Supplier No. 466000
Only null left
Add to Cart
Add to Cart
Not a toy. Product intended for ages 13 and up.
Includes
Includes meter stick, object marker and screen, screen support, metal stick supports, lens holder
A simple and inexpensive apparatus to help demonstrate basic optics principles. The GSC Optical Bench provides an accurate demonstration of basic optics principles.
  • Simple form optical bench
  • Includes instructions
Specifications
Includes With meter stick
For Use With (Application) For demonstration of basic optics principles
Length (Metric) Focal 100 cm
Product Type Optical Bench
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov