Learn More
GSC Go Science Crazy Precision Gyroscope
SCI_ED
Catalog No. S13854 Shop All GSC Products
Click to view available options
Precision, gyroscope
Catalog No.
S13854 Precision, gyroscope
1 options
Not a toy. Product intended for ages 13 and up.
Catalog No. S13854 Supplier GSC Go Science Crazy Supplier No. 14006
Not a toy. Product intended for ages 13 and up.
Includes
Plastic gimbals, rocker and pivoting housing, gyroscope, and cord
A fun and educational apparatus that demonstrates the properties of gyroscopes. The GSC Precision Gyroscope is a fascinating mechanical device that seems to defy the laws of gravity. This is a well-balanced, high quality gyroscope with adjustable bearings. The unit includes a plastic gimbals, rocker and pivoting housing, the gyroscope and cord.
  • Precision gyroscope
  • Well balanced
  • High quality
  • Adjustable bearings
Specifications
Product Type Gyroscope
For Use With (Application) Demonstrates why gyroscopes resist changes in direction when spinning
Includes Gyroscope with plastic gimbal housing; Plastic handle and cord; Two gyroscope cradles, one rocking and one pivoting; Six washers (three large, three small); S-hook
Dimensions (L x W x H) 4.44 x 4.25 x 2.22 in.
Weight (English) 10 oz.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov