Learn More
GSC Go Science Crazy Pressure Globe / Harbottle Apparatus
SCI_ED
Catalog No. S13855 Shop All GSC Products
Click to view available options
Includes:
Harbottle, balloon, and rubber stopper
Catalog No. Includes
S13855 Harbottle, balloon, and rubber stopper
1 options
This item is not returnable. View return policy
Not a toy. Product intended for ages 13 and up. May Contain Latex!
Catalog No. S13855 Supplier GSC Go Science Crazy Supplier No. 791
This item is not returnable. View return policy
Not a toy. Product intended for ages 13 and up. May Contain Latex!
Includes
Harbottle, Rubber Stopper and Balloon
Helps students to understand the meaning of fluid pressure and pressure reduction. The GSC Pressure Globe / Harbottle Apparatus helps students to understand the meaning of fluid pressure and pressure reduction. Simply place a balloon into the opening of the glass Harbottle and stretch the mouth of the balloon over the mouth of the bottle.
  1. Place a balloon into opening of glass Harbottle and stretch mouth of balloon over mouth of bottle
  2. Blow into balloon and when fully inflated, stopper bottom hole of Harbottle
  3. Balloon remains inflated although open to atmosphere
  • Great for teaching about pressure
  • Includes balloon and stopper
  • Harbottle made from glass
Specifications
Product Type Harbottle apparatus
Includes Harbottle, balloon, and rubber stopper
Material Glass, Rubber
For Use With (Application) For discussion on pressure, vacuum, pumps, and weather