Learn More
GSC Go Science Crazy Resonance Apparatus
SCI_ED
Catalog No. S13857 Shop All GSC Products
Click to view available options
For Use With (Application):
Measures velocity of sound in air
Catalog No. For Use With (Application)
S13857 Measures velocity of sound in air
1 options
Not a toy. Product intended for ages 13 and up. May Contain Latex!
Catalog No. S13857 Supplier GSC Go Science Crazy Supplier No. 418061
Not a toy. Product intended for ages 13 and up. May Contain Latex!
A simple apparatus that measures the velocity of sound in the air. The GSC Resonance Apparatus measures the velocity of sound in the air. Partially fill the resonance tube with water and then sound a tuning fork of known frequency at the tubes open upper end of the Resonance Tube.
  • Easily measure the velocity of sound
  • Includes instructions
Specifications
Product Type Resonance Apparatus
Material Glass (Tube), Cast Iron (Support Stand)
For Use With (Application) Measures velocity of sound in air
Includes Instructions
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov