Learn More
GSC Go Science Crazy Second Law of Motion
SCI_ED
Catalog No. S13619 Shop All GSC Products
Click to view available options
Dimensions (L x W x H):
12 x 4.5 x 2 in.
Catalog No. Dimensions (L x W x H)
S13619 12 x 4.5 x 2 in.
1 options
Not a toy. Product intended for ages 13 and up.
Catalog No. S13619 Supplier GSC Go Science Crazy Supplier No. 1476
Not a toy. Product intended for ages 13 and up.
A great for unit for students to easily visualize and grasp Newton's Second Law of Motion. The GSC Second Law of Motion helps students to visualize and grasp Newton's Second Law of Motion. This sturdy unit demonstrates that a horizontally projected ball and a vertically dropped ball land at the same time.
  • Small and easy to use unit
  • Easy visual demonstration
  • Can clamp to a table or stand
  • Instructions included
Specifications
For Use With (Application) Demonstrates Newton second law of motion
Includes 2 steel balls, cast iron rod, and instructions
Dimensions (L x W x H) 12 x 4.5 x 2 in.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov