Learn More
GSC Go Science Crazy Refractive Tank Semi Circular
SCI_ED
Catalog No. S13673 Shop All GSC Products
Click to view available options
Quantity:
12 Pk.
Catalog No. Quantity
S13673 12 Pk.
1 options
Not a toy. Product intended for ages 13 and up.
Catalog No. S13673 Supplier GSC Go Science Crazy Supplier No. SCRD10
Not a toy. Product intended for ages 13 and up.
Easily demonstrate Snell's law and show how light slows and refracts by entering a fluid. The GSC Refractive Tank Semi Circular, Pack 12 demonstrates Snell’s Law, which is what happens when light moves from air space across a boundary into a fluid at various angles.
  • Visually demonstrate Snell's Law
  • Can be filled with various fluids
  • Semi-circular design
  • Comes in a 12 pack
Specifications
Quantity 12 Pk.
For Use With (Application) Demonstrate Snell's Law
Product Type Refractive Tank
Shape Semi-circular
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov