Learn More
GSC Go Science Crazy Stepped Pulley Wheel & Axle
SCI_ED
Catalog No. S13859 Shop All GSC Products
Click to view available options
Pulley, stepped, wheel & axle
Catalog No.
S13859 Pulley, stepped, wheel & axle
1 options
Not a toy. Product intended for ages 13 and up.
Catalog No. S13859 Supplier GSC Go Science Crazy Supplier No. 2302
Not a toy. Product intended for ages 13 and up.
A great teaching tool for demonstrating the conservation of energy. The GSC Stepped Pulley Wheel & Axle is often used to demonstrate principals of conservation of energy and simple machines. The pulley is constructed of plastic with a steel rod and roller bearing hub.
  • Great for learning or practical use
  • Four different pulley sizes
  • Works great with a stand or clamp
Specifications
Product Type Pulley, Stepped with Axle
Description Step sizes: 1.5, 2, 2.5 and 4 in.
Material Plastic, Steel (rod and roller bearing hub)
For Use With (Application) Demonstrate principals of conservation of energy and simple machines
For Use With (Equipment) Support stand, Clamp
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov