Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSC Go Science Crazy Support Ring Clamp
SCI_ED

Catalog No. S13757 Shop All GSC Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Diameter (English):
2 in. (Outer Dia.)
3 in. (Outer Dia.)
4 in. (Outer Dia.)
5 in. (Outer Dia.)
6 in. (Outer Dia.)
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (English)
S13757 6 in. (Outer Dia.)
S13753 2 in. (Outer Dia.)
S13754 3 in. (Outer Dia.)
S13755 4 in. (Outer Dia.)
S13756 5 in. (Outer Dia.)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options

Not a toy. Product intended for ages 13 and up.

Catalog No. S13757 Supplier GSC Go Science Crazy Supplier No. 4SRC4410
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Not a toy. Product intended for ages 13 and up.

Great for holding all sorts of lab equipment in a small space. Works best with GSC Support Stands and Rods. The GSC Support Ring Clamp is a cast-iron clamp with nickel plated steel set screws. Can be clamped to a support stand rod for holding lab equipment.

  • Clamp made from cast iron
  • Nickel-plated steel screws
  • Great for support stands/rods

Specifications

Diameter (English) 6 in. (Outer Dia.)
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.