Learn More
GSC Go Science Crazy Support Stand with Rings
SCI_ED
Catalog No. S13752 Shop All GSC Products
Encompass
Change view
Click to view available options
Product Type:
2-Ring
3-Ring
4-Ring
5-Ring
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S13752 5-Ring
S13749 2-Ring
S13750 3-Ring
S13751 4-Ring
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Not a toy. Product intended for ages 13 and up.
Catalog No. S13752 Supplier GSC Go Science Crazy Supplier No. 4SSC17R5
This item is not returnable. View return policy
Not a toy. Product intended for ages 13 and up.
Includes
Includes cast metal support rings with zinc finish
Great for holding all sorts of lab equipment in a small space. The GSC Support Stand with Rings is made from cast iron, with an enamel finish that is acid resistant. Includes a zinc plated steel rod that screws into the base support. Supplied with support rings which are also cast-metal with a zinc finish.
  • Stand made from cast iron
  • Acid resistant
  • Plated steel support rod
  • Rod threaded to fit base
  • Includes support rings
Specifications
Product Type 5-Ring
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov