Learn More
GSC Go Science Crazy Wave Demonstration Kit
SCI_ED
Catalog No. S13646 Shop All GSC Products
Click to view available options
Wave demonstration, kit
Catalog No.
S13646 Wave demonstration, kit
1 options
Not a toy. Product intended for ages 13 and up. May Contain Latex!
Catalog No. S13646 Supplier GSC Go Science Crazy Supplier No. SLINKYKT
Not a toy. Product intended for ages 13 and up. May Contain Latex!
A welcome addition to any classroom. This Kit will help students visualize how waves propagation works. The GSC Wave Demonstration Kit comes with one long, narrow steel helical spring and, one larger helical spring that are matched to give nearly equal amplitudes of reflected and transmitted pulses and waves. A string and instructions are also included in the kit.
  • Kit includes 2 springs and a string
  • Easily visualize wave propagation
Specifications
Product Type Kit, Wave Demonstration
Includes One long, narrow steel spring; One larger helical spring; String
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov