Learn More
Aviva Systems Biology Corp GSCL Antibody - C-terminal region (ARP38599_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38599_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against GSCL. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human GSCL. Alternate names: GSCLApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53109