Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gold Standard Diagnostics GSD RPR Diluent
SDP

Catalog No. 23700137
Change view
Click to view available options
Disposable:
Single-use
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Disposable
23-700-137 Single-use
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-700-137 Supplier Gold Standard Diagnostics Supplier No. GSD011600DIL
Add to Cart
Add to Cart

New diluent used with the AIX1000 2.1 for high to extra high titer testing and the fully automated RPR for Syphilis Serology

  • For use with high titers 1:32 to 1:256, extra high titers 1:512 to 1:2048

Specifications

CE Marker Yes
Clia Complexity Moderate (Serum, Plasma or Whole Blood), Waived (Other)
Detectable Analytes None
Type RPR Diluent
Disposable Single-use
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.