Learn More
Biomatik Corporation Gsdma Polyclonal Antibody
Supplier:  Biomatik Corporation CAC11466
Encompass_Preferred
Gsdma Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to GSDMA. The Gsdma Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Gasdermin-A Protein (64-172Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: GSDMA. Accession Number: Q96QA5. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6769