Learn More
ABclonal Technology GSDMC Rabbit pAb
SDP
Supplier:  ABclonal Technology A145501000UL
Encompass_Preferred
Predicted to enable phosphatidylinositol-4,5-bisphosphate binding activity, phosphatidylinositol-4-phosphate binding activity, and phosphatidylserine binding activity. Predicted to be involved in defense response to bacterium and pyroptosis. Predicted to act upstream of or within intestinal epithelial cell development. Located in cytoplasm.
Catalog No. 50-001-08591