Learn More
MyBioSource Human Gasdermin D (GSDMD) ELISA Kit
SDP
Supplier:  MyBioSource MBS913704396TESTS
Encompass_Preferred
The MyBioSource Human Gasdermin D ELISA Kit quantitatively measures Gasdermin D in various biological fluids using a sandwich enzyme immunoassay. It involves a monoclonal antibody-coated 96-well plate, biotin-conjugated polyclonal antibody, and HRP with a TMB substrate that changes color to indicate Gasdermin D levels, measured at 450 nm.
Catalog No. NC2386577