Learn More
MSC GSG Micrometer and Caliper Calibration Kit, 1-12 in, 0.0001 in Accuracy
Catalog No. 5000528647
Encompass
This calibration kit is designed for the verification of accuracy for lab and shop measuring tools, ensuring correct measurements and preventing the use of worn tools on the manufacturing floor. It provides 0.0001 in accuracy and features chrome-plated gage surfaces for extra longevity, held to a tolerance of ±0.0001 in.
  • Accurate to 0.0001 in.
  • Verifies accuracy of micrometers, calipers, and variable snap gages.
  • Supports calibration for a 1-12 in measurement range.
  • Gage surfaces are chrome plated for longevity and held to a tolerance of ±0.0001 in.
  • Manufactured in the United States.
  • Case not included.
Catalog No. 50-005-28647 Supplier MSC Supplier No. 35115120
This calibration kit is designed for the verification of accuracy for lab and shop measuring tools, ensuring correct measurements and preventing the use of worn tools on the manufacturing floor. It provides 0.0001 in accuracy and features chrome-plated gage surfaces for extra longevity, held to a tolerance of ±0.0001 in.
  • Accurate to 0.0001 in.
  • Verifies accuracy of micrometers, calipers, and variable snap gages.
  • Supports calibration for a 1-12 in measurement range.
  • Gage surfaces are chrome plated for longevity and held to a tolerance of ±0.0001 in.
  • Manufactured in the United States.
  • Case not included.