Learn More
Cayman Chemical GsG Stndard 1ea
SDP
Catalog No. 502895166
Encompass_Preferred
An FXR agonist induces luminescence in a reporter assay using HEK293T cells expressing human FXR (EC50 18 nM) decreases the serum levels of ALT and total cholesterol as well as hepatic inflammation fat levels and fibrosis in a mouse model of NASH induced by a high-fat diet and CCl4 at 1 3 or 10 mg/kg per day
Catalog No. 50-289-5166 Supplier Cayman Chemical Supplier No. 7030141 EA
An FXR agonist induces luminescence in a reporter assay using HEK293T cells expressing human FXR (EC50 18 nM) decreases the serum levels of ALT and total cholesterol as well as hepatic inflammation fat levels and fibrosis in a mouse model of NASH induced by a high-fat diet and CCl4 at 1 3 or 10 mg/kg per day