Learn More
Aviva Systems Biology Corp HASPIN Antibody - middle region (ARP60553_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60553_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GSG2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human GSG2. Alternate names: HASPINApplicationWB. Species reactivity: Human
Catalog No. 50-287-082