Learn More
Apexbio Technology LLC GSH/GSSG RATIO FLU KIT-200mg
SDP
Catalog No. 5000567727
Encompass_Preferred
GSH/GSSG Ratio Assay Kit (Fluorometric) is a vital tool in oxidative stress and redox biology research assessing the balance between reduced glutathione (GSH) and its oxidized form (GSSG) Originating from studies on cellular oxidative mechanisms it targets pathways involved in maintaining redox homeostasis critical for cellular function and survival Often used in vitro at nanomolar to low micromolar concentrations this kit supports various applications including cell model experiments animal studies and drug screenings Experimental concentrations typically depend on the specific research design The quantification of the GSH/GSSG ratio offers insights into cellular oxidative stress levels aiding in the understanding of pathologies such as cancer and neurodegenerative diseases
Catalog No. 50-005-67727 Supplier Apexbio Technology LLC Supplier No. K2109200
Add to Cart
Add to Cart
GSH/GSSG Ratio Assay Kit (Fluorometric) is a vital tool in oxidative stress and redox biology research assessing the balance between reduced glutathione (GSH) and its oxidized form (GSSG) Originating from studies on cellular oxidative mechanisms it targets pathways involved in maintaining redox homeostasis critical for cellular function and survival Often used in vitro at nanomolar to low micromolar concentrations this kit supports various applications including cell model experiments animal studies and drug screenings Experimental concentrations typically depend on the specific research design The quantification of the GSH/GSSG ratio offers insights into cellular oxidative stress levels aiding in the understanding of pathologies such as cancer and neurodegenerative diseases