Learn More
AdipoGen GSH-OEt
SDP
Catalog No. 502560845
Encompass_Preferred
Chemical. CAS 92614-59-0. Formula C12H21N3O6S. MW 335.38. Synthetic. GSH-OEt is cellpermeable and has been used to protect cells against radiation damage, oxidants and various toxic compounds including heavy metals. GSH-OEt is a protective agent against cellular damage, such as cataracts and mitochondrial degeneration. It undergoes hydrolysis by intracellular esterases thereby increasing intracellular GSH concentration in many tissues and cell types. Glutathione monoethyl ester may be used to supplement cellular pools of GSH in vitro and in vivo and can prevent ROS formation, neutralize toxic products and block apoptosis pathway. The effect of glutathione GSH and glutathione ethyl ester GSH-OEt supplementation on GSH homeostasis and exercise-induced oxidative stress was also examined. Glutathione ethyl ester protects against cisplatin-induced ototoxicity in the rat.
Catalog No. 50-256-0845 Supplier AdipoGen Supplier No. CDXG0006M500
Chemical. CAS 92614-59-0. Formula C12H21N3O6S. MW 335.38. Synthetic. GSH-OEt is cellpermeable and has been used to protect cells against radiation damage, oxidants and various toxic compounds including heavy metals. GSH-OEt is a protective agent against cellular damage, such as cataracts and mitochondrial degeneration. It undergoes hydrolysis by intracellular esterases thereby increasing intracellular GSH concentration in many tissues and cell types. Glutathione monoethyl ester may be used to supplement cellular pools of GSH in vitro and in vivo and can prevent ROS formation, neutralize toxic products and block apoptosis pathway. The effect of glutathione GSH and glutathione ethyl ester GSH-OEt supplementation on GSH homeostasis and exercise-induced oxidative stress was also examined. Glutathione ethyl ester protects against cisplatin-induced ototoxicity in the rat.