Learn More
AdipoGen GSH-Oet, 92614-59-0, MFCD00156117, 100 mg
SDP
Supplier:  AdipoGen CDXG0006M100
Encompass_Preferred
MF: C12H21N3O6S, MW: 335.38, ≥90%, Appearance: Solid. GSH-OEt is cellpermeable and has been used to protect cells against radiation damage, oxidants and various toxic compounds including heavy metals. GSH-OEt is a protective agent against cellular damage, such as cataracts and mitochondrial degeneration.
Catalog No. 50-596-632