Learn More
Aviva Systems Biology Corp GSH2 Antibody - C-terminal region (ARP39863_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39863_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against GSH2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human GSH2. Alternate names: GSH2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57208