Learn More
Aviva Systems Biology Corp GSH2 Antibody - N-terminal region (ARP32208_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32208_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GSH2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human GSH2. Alternate names: GSH2ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-52855