Learn More
TARGETMOL CHEMICALS INC GSK-1482160 5MG
SDP
Catalog No. 502579559
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. GSK-1482160 is a CNS-penetrant negative allosteric modulator of P2X7 receptor with oral activity. The P2X7 receptor is involved in the production of pro-inflammatory cytokines by central and peripheral immune cells so GSK-1482160 has the potential to study inflammatory diseases. GSK-1482160 shows excellent in vitro potency (in functional and electrophysiological assays at recombinant and naive P2X7 receptors across multiple species including humans) and cross-target selectivity. purity: 99%
Catalog No. 50-257-9559 Supplier TARGETMOL CHEMICALS INC Supplier No. T610255MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. GSK-1482160 is a CNS-penetrant negative allosteric modulator of P2X7 receptor with oral activity. The P2X7 receptor is involved in the production of pro-inflammatory cytokines by central and peripheral immune cells so GSK-1482160 has the potential to study inflammatory diseases. GSK-1482160 shows excellent in vitro potency (in functional and electrophysiological assays at recombinant and naive P2X7 receptors across multiple species including humans) and cross-target selectivity. purity: 99%