Learn More
Medchemexpress LLC GSK-1520489A | 1042433-41-9 | 99.8% | 425.50 | 5 MG
SDP
Catalog No. 5000359354
Encompass_Preferred
GSK-1520489A is an inhibitor of PKMYT1 (active protein kinase) with an IC50 of 115 nM and a Ki of 10.94 nM. It is intended for research use only.
  • Inhibits phosphorylation of Tyr15 in Cdk1 at a concentration of 4 μM after 24 hours of treatment.
  • Reduces Cdk1-phosphorylation at tyrosine in HT29 cells when treated with 4 μM for 24 hours.
Catalog No. 50-003-59354 Supplier Medchemexpress LLC Supplier No. HY1473125MG
GSK-1520489A is an inhibitor of PKMYT1 (active protein kinase) with an IC50 of 115 nM and a Ki of 10.94 nM. It is intended for research use only.
  • Inhibits phosphorylation of Tyr15 in Cdk1 at a concentration of 4 μM after 24 hours of treatment.
  • Reduces Cdk1-phosphorylation at tyrosine in HT29 cells when treated with 4 μM for 24 hours.