Learn More
Medchemexpress LLC Gsk-1520489a | 1042433-41-9 | 99.8% | C21H23N5O3S | 10MG
SDP
Catalog No. 5000259527
Encompass_Preferred
GSK-1520489A is a research-grade small-molecule inhibitor of PKMYT1 (protein kinase, membrane associated tyrosine/threonine 1). It demonstrates biochemical potency against PKMYT1 (IC50 = 115 nM; Ki = 10.94 nM) and is supplied as a high-purity solid for biochemical and cell-based research. CAS 1042433-41-9; molecular weight 425.50; formula C21H23N5O3S.
  • Potent PKMYT1 inhibition (IC50 = 115 nM; Ki = 10.94 nM).
  • High purity suitable for research (99.8%).
  • Supplied as a solid with recommended storage conditions.
  • Suitable for biochemical kinase assays and cell-based studies.
  • Well-characterized molecular properties: formula C21H23N5O3S, MW 425.50.
  • A datasheet provides analytical and handling information.
Catalog No. 50-002-59527 Supplier Medchemexpress LLC Supplier No. HY14731210MG
GSK-1520489A is a research-grade small-molecule inhibitor of PKMYT1 (protein kinase, membrane associated tyrosine/threonine 1). It demonstrates biochemical potency against PKMYT1 (IC50 = 115 nM; Ki = 10.94 nM) and is supplied as a high-purity solid for biochemical and cell-based research. CAS 1042433-41-9; molecular weight 425.50; formula C21H23N5O3S.
  • Potent PKMYT1 inhibition (IC50 = 115 nM; Ki = 10.94 nM).
  • High purity suitable for research (99.8%).
  • Supplied as a solid with recommended storage conditions.
  • Suitable for biochemical kinase assays and cell-based studies.
  • Well-characterized molecular properties: formula C21H23N5O3S, MW 425.50.
  • A datasheet provides analytical and handling information.