Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GSK 1562590 hydrochloride | 1003878-07-6 | 99.0% | 617.95 | 10 MG
SDP

Catalog No. 5000346788
Encompass_Preferred

GSK 1562590 hydrochloride is a high-affinity and selective antagonist of the urotensin-II receptor (UT), with pKis ranging from 9.14 to 9.66 for mammalian recombinant (mouse, rat, cat, monkey, human) and native UT. It is intended for research use only.

  • High affinity and selective antagonist of urotensin-II receptor (UT).
  • PKis of 9.14-9.66 for mammalian recombinant (mouse, rat, cat, monkey, human) and native UT.
  • Appears as a solid, white to off-white in color.
  • Soluble in DMSO (25 mg/mL).
  • Storage recommendations: -20°C, protect from light, stored under nitrogen; in solvent: -80°C for 6 months; -20°C for 1 month.

Catalog No. 50-003-46788 Supplier Medchemexpress LLC Supplier No. HY10844650MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GSK 1562590 hydrochloride is a high-affinity and selective antagonist of the urotensin-II receptor (UT), with pKis ranging from 9.14 to 9.66 for mammalian recombinant (mouse, rat, cat, monkey, human) and native UT. It is intended for research use only.

  • High affinity and selective antagonist of urotensin-II receptor (UT).
  • PKis of 9.14-9.66 for mammalian recombinant (mouse, rat, cat, monkey, human) and native UT.
  • Appears as a solid, white to off-white in color.
  • Soluble in DMSO (25 mg/mL).
  • Storage recommendations: -20°C, protect from light, stored under nitrogen; in solvent: -80°C for 6 months; -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.