Learn More
Medchemexpress LLC GSK 1562590 (hydrochloride) | 1003878-07-6 | 99.0% | 617.95 | 50 MG
SDP
Catalog No. 5000346792
Encompass_Preferred
GSK 1562590 hydrochloride is a high affinity and selective antagonist of urotensin-II receptor (UT), with pKis of 9.14-9.66 for mammalian recombinant (mouse, rat, cat, monkey, human) and native UT.
  • High affinity and selective antagonist of urotensin-II receptor (UT).
Catalog No. 50-003-46792 Supplier Medchemexpress LLC Supplier No. HY10844610MG
GSK 1562590 hydrochloride is a high affinity and selective antagonist of urotensin-II receptor (UT), with pKis of 9.14-9.66 for mammalian recombinant (mouse, rat, cat, monkey, human) and native UT.
  • High affinity and selective antagonist of urotensin-II receptor (UT).