Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GSK-2256098 hydrochloride | 1416771-10-2 | 99.9% | 451.35 | C20H24Cl2N6O2 | 25 MG
SDP

Catalog No. 5000214810
Encompass_Preferred

GSK-2256098 hydrochloride is the hydrochloride salt of a selective focal adhesion kinase (FAK) inhibitor supplied for research use to study cell adhesion, migration, proliferation, angiogenesis, and tumor cell survival. It is intended for in vitro research only and is reported at high purity.

  • Selective focal adhesion kinase (FAK) inhibitor suitable for in vitro studies.
  • High reported purity, 99.9%.
  • CAS number 1416771-10-2 for unambiguous identification.
  • Molecular formula C20H24Cl2N6O2 and molecular weight 451.35 g/mol.
  • Available as a 25 mg research-grade package.

Catalog No. 50-002-14810 Supplier Medchemexpress LLC Supplier No. HY100498A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

GSK-2256098 hydrochloride is the hydrochloride salt of a selective focal adhesion kinase (FAK) inhibitor supplied for research use to study cell adhesion, migration, proliferation, angiogenesis, and tumor cell survival. It is intended for in vitro research only and is reported at high purity.

  • Selective focal adhesion kinase (FAK) inhibitor suitable for in vitro studies.
  • High reported purity, 99.9%.
  • CAS number 1416771-10-2 for unambiguous identification.
  • Molecular formula C20H24Cl2N6O2 and molecular weight 451.35 g/mol.
  • Available as a 25 mg research-grade package.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.