Learn More
Medchemexpress LLC HY-14362 5mg Medchemexpress, GSK-25 CAS:874119-56-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY143625MG
Encompass_Preferred
Medchemexpress, HY-14362 5mg GSK-25 CAS:874119-56-9 GSK-25 is a potent, selective and orally bioavailable ROCK1 inhibitor (IC50=7 nM). GSK-25 maintains good selectivity against a panel of 31 kinases (>100 fold), as well as RSK1 and p70S6K (RSK1: IC50=398 nM, p70S6K: IC50=1 μM). GSK-25 inhibits P450 profile (IC50s of 2.5, 5.2, 2.5 µM for CYP2C9, CYP2D6, CYP3A4, respectively). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9901