Learn More
Medchemexpress LLC GSK-2793660 | 1613458-78-8 | 99.9% | 393.91 | 50 MG
SDP
Catalog No. 5000435734
Encompass_Preferred
GSK-2793660 is an orally active and irreversible inhibitor of Cathepsin C (CTSC). It is intended for laboratory chemical use by experienced personnel in the research of conditions such as bronchiectasis, cystic fibrosis, and anti-neutrophil cytoplasmic autoantibody (ANCA)-associated vasculitis.
  • Orally active and irreversible inhibitor
  • Targets Cathepsin C (CTSC)
  • Supports research of bronchiectasis
  • Supports research of cystic fibrosis
  • Supports research of anti-neutrophil cytoplasmic autoantibody (ANCA)-associated vasculitis
Catalog No. 50-004-35734 Supplier Medchemexpress LLC Supplier No. HY112318A50MG
GSK-2793660 is an orally active and irreversible inhibitor of Cathepsin C (CTSC). It is intended for laboratory chemical use by experienced personnel in the research of conditions such as bronchiectasis, cystic fibrosis, and anti-neutrophil cytoplasmic autoantibody (ANCA)-associated vasculitis.
  • Orally active and irreversible inhibitor
  • Targets Cathepsin C (CTSC)
  • Supports research of bronchiectasis
  • Supports research of cystic fibrosis
  • Supports research of anti-neutrophil cytoplasmic autoantibody (ANCA)-associated vasculitis