Learn More
Medchemexpress LLC Gsk-2793660 free base | 1613458-70-0 | 357.45 | 10 MG
SDP
Catalog No. 5000438375
Encompass_Preferred
GSK-2793660 (free base) is an oral, irreversible inhibitor of Cathepsin C (CTSC) and can be used for the research of bronchiectasis. At equivalent molar concentrations, both the salt and free forms of a compound exhibit comparable biological activity, however, the salt form usually boasts enhanced water solubility and stability. It is intended for research use only.
  • Oral, irreversible inhibitor of Cathepsin C (CTSC).
  • Useful for research related to bronchiectasis, cystic fibrosis, non-cystic fibrosis bronchiectasis, and anti-neutrophil cytoplasmic autoantibody (ANCA)-associated vasculitis.
  • Free base form offers comparable biological activity to the salt form.
Catalog No. 50-004-38375 Supplier Medchemexpress LLC Supplier No. HY112318A10MG
GSK-2793660 (free base) is an oral, irreversible inhibitor of Cathepsin C (CTSC) and can be used for the research of bronchiectasis. At equivalent molar concentrations, both the salt and free forms of a compound exhibit comparable biological activity, however, the salt form usually boasts enhanced water solubility and stability. It is intended for research use only.
  • Oral, irreversible inhibitor of Cathepsin C (CTSC).
  • Useful for research related to bronchiectasis, cystic fibrosis, non-cystic fibrosis bronchiectasis, and anti-neutrophil cytoplasmic autoantibody (ANCA)-associated vasculitis.
  • Free base form offers comparable biological activity to the salt form.