Learn More
Apexbio Technology LLC GSK 2830371 1404456-53-6 10mg
SDP
Catalog No. 5000090962
Encompass_Preferred
GSK 2830371 (CAS 1404456-53-6) is a potent orally bioavailable small molecule that acts as an allosteric inhibitor of Wip1 phosphatase By selectively targeting Wip1 a serine/threonine phosphatase involved in the regulation of DNA damage response pathways GSK 2830371 disrupts downstream signaling events critical for cell cycle progression and genomic stability The compound exhibits an IC of 6 nM against Wip1 in biochemical assays reflecting high inhibitory potency GSK 2830371 is utilized in biomedical research to investigate Wip1 s role in oncogenic processes and to explore the therapeutic potential of Wip1 inhibition in cancer and related pathologies
Catalog No. 50-000-90962 Supplier Apexbio Technology LLC Supplier No. B116910
GSK 2830371 (CAS 1404456-53-6) is a potent orally bioavailable small molecule that acts as an allosteric inhibitor of Wip1 phosphatase By selectively targeting Wip1 a serine/threonine phosphatase involved in the regulation of DNA damage response pathways GSK 2830371 disrupts downstream signaling events critical for cell cycle progression and genomic stability The compound exhibits an IC of 6 nM against Wip1 in biochemical assays reflecting high inhibitory potency GSK 2830371 is utilized in biomedical research to investigate Wip1 s role in oncogenic processes and to explore the therapeutic potential of Wip1 inhibition in cancer and related pathologies