Learn More
Apexbio Technology LLC GSK 2837808A 1445879-21-9 25mg
SDP
Catalog No. 5000089456
Encompass_Preferred
GSK 2837808A (CAS 1445879-21-9) is a small-molecule inhibitor that selectively targets lactate dehydrogenase A (LDHA) exhibiting potent inhibitory activity with IC50 values of 1 9 nM for LDHA and 14 nM for LDHB In cellular studies GSK 2837808A at 10 M reduced glucose consumption in Snu398 hepatocellular carcinoma cells but not in HepG2 cells indicating cell line-specific metabolic effects Pharmacokinetic analysis in rodents demonstrated rapid clearance following intravenous administration and minimal systemic exposure after oral dosing GSK 2837808A serves as a valuable tool for investigating LDHA function and metabolic regulation in cancer research
Catalog No. 50-000-89456 Supplier Apexbio Technology LLC Supplier No. B492925
GSK 2837808A (CAS 1445879-21-9) is a small-molecule inhibitor that selectively targets lactate dehydrogenase A (LDHA) exhibiting potent inhibitory activity with IC50 values of 1 9 nM for LDHA and 14 nM for LDHB In cellular studies GSK 2837808A at 10 M reduced glucose consumption in Snu398 hepatocellular carcinoma cells but not in HepG2 cells indicating cell line-specific metabolic effects Pharmacokinetic analysis in rodents demonstrated rapid clearance following intravenous administration and minimal systemic exposure after oral dosing GSK 2837808A serves as a valuable tool for investigating LDHA function and metabolic regulation in cancer research