Learn More
Abcam GSK-2982772, 1MG
Supplier:  Abcam AB2869621MG
Encompass_Preferred
MW 377.4 Da, Purity >=98%. A potent and ATP-competitive receptor-interacting protein-1 (RIP1) kinase inhibitor (IC₅₀ = 1 nM).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-220-5806