Learn More
Medchemexpress LLC GSK-3 beta protein, human (P. pastoris, His) | >90.0% | 100UG
SDP
Catalog No. 5000176939
Encompass_Preferred
Recombinant human glycogen synthase kinase-3 beta (GSK-3β) is supplied as a full-length, His-tagged protein expressed in Pichia pastoris. The protein is an active serine-threonine kinase involved in glucose homeostasis and Wnt signaling and is suitable for kinase assays, biochemical characterization, and structural studies.
  • Full-length human isoform-1
  • Expressed in Pichia pastoris for eukaryotic post-translational modifications
  • N-6xHis tag enables affinity purification and detection
  • Purity >90.0% as determined by reducing SDS-PAGE
  • Approx. 50 kDa molecular weight
  • Suitable for kinase assays and biochemical studies
Catalog No. 50-001-76939 Supplier Medchemexpress LLC Supplier No. HYP700591100UG
Recombinant human glycogen synthase kinase-3 beta (GSK-3β) is supplied as a full-length, His-tagged protein expressed in Pichia pastoris. The protein is an active serine-threonine kinase involved in glucose homeostasis and Wnt signaling and is suitable for kinase assays, biochemical characterization, and structural studies.
  • Full-length human isoform-1
  • Expressed in Pichia pastoris for eukaryotic post-translational modifications
  • N-6xHis tag enables affinity purification and detection
  • Purity >90.0% as determined by reducing SDS-PAGE
  • Approx. 50 kDa molecular weight
  • Suitable for kinase assays and biochemical studies