Learn More
Medchemexpress LLC Glycogen synthase kinase-3 beta | 443900-95-6 | MFCD00285876 | >90.0% | 50UG
SDP
Catalog No. 5000176940
Encompass_Preferred
Recombinant human glycogen synthase kinase-3 beta (GSK-3β) expressed in Pichia pastoris and supplied as an N-6xHis-tagged, active serine/threonine kinase for use in biochemical assays and research applications. The product is characterized for purity and delivered in small vial sizes appropriate for enzymatic studies.
Catalog No. 50-001-76940 Supplier Medchemexpress LLC Supplier No. HYP70059150UG
Recombinant human glycogen synthase kinase-3 beta (GSK-3β) expressed in Pichia pastoris and supplied as an N-6xHis-tagged, active serine/threonine kinase for use in biochemical assays and research applications. The product is characterized for purity and delivered in small vial sizes appropriate for enzymatic studies.