Learn More
Medchemexpress LLC GSK-3β inhibitor 3 | 1448990-73-5 | 99.2% | 327.37 | C18H14FNO2S | 5 MG
SDP
Catalog No. 5000222601
Encompass_Preferred
GSK-3β inhibitor 3 is a potent, selective, irreversible covalent inhibitor of glycogen synthase kinase 3β (GSK-3β) intended for in vitro research applications. Reported IC50 is 6.6 μM. The compound is supplied as a solid and as a 10 mM solution in DMSO for laboratory use.
  • Potent, selective inhibition of GSK-3β.
  • Irreversible, covalent binding mechanism.
  • Reported IC50 of 6.6 μM for biochemical assays.
  • Supplied as solid and as a 10 mM solution in DMSO for convenience.
  • High purity suitable for research (≈99.2%).
Catalog No. 50-002-22601 Supplier Medchemexpress LLC Supplier No. HY1414805MG
GSK-3β inhibitor 3 is a potent, selective, irreversible covalent inhibitor of glycogen synthase kinase 3β (GSK-3β) intended for in vitro research applications. Reported IC50 is 6.6 μM. The compound is supplied as a solid and as a 10 mM solution in DMSO for laboratory use.
  • Potent, selective inhibition of GSK-3β.
  • Irreversible, covalent binding mechanism.
  • Reported IC50 of 6.6 μM for biochemical assays.
  • Supplied as solid and as a 10 mM solution in DMSO for convenience.
  • High purity suitable for research (≈99.2%).