Learn More
Apexbio Technology LLC GSK-3 Inhibitor IX (BIO), 10mg. Cas: 667463-62-9 MFCD: MFCD08705318. GSK-3α/GSK-3β inhibitor, cell-permeable, ATP-competitive and reversible
SDP
Supplier:  Apexbio Technology LLC B153810
Encompass_Preferred
GSK-3 inhibitor IX (BIO) is a potent, selective, cell-permeable, ATP-competitive and reversible inhibitor of GSK-3 (glycogen synthase kinase-3) and GSK-3 (lycogen synthase kinase-3) (IC50 = 5nM for GSK-3). Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5253