Learn More
Enzo Life Sciences GSK-3 inhibitor XIII (1mg). CAS: 404828-08-6
SDP
Catalog No. 502013411
Encompass_Preferred
Alternative name: (5-Methyl-1H-pyrazol-3-yl)-(2-phenylquinazolin-4-yl)amine. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -80°C.
Catalog No. 50-201-3411 Supplier Enzo Life Sciences Supplier No. ENZCHM1240001
Alternative name: (5-Methyl-1H-pyrazol-3-yl)-(2-phenylquinazolin-4-yl)amine. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -80°C.