Learn More
Medchemexpress LLC Gsk-3 inhibitor xiii | 404828-14-4 | 98.79% | 301.35 | 10 MG
SDP
Catalog No. 5000438414
Encompass_Preferred
GSK-3 Inhibitor XIII is a potent and ATP-competitive GSK-3 inhibitor with a Ki of 24 nM.
  • Potent and ATP-competitive GSK-3 inhibitor
  • Ki of 24 nM
  • White to off-white solid appearance
  • High purity of 98.79%
  • Soluble in DMSO (100 mg/mL)
Catalog No. 50-004-38414 Supplier Medchemexpress LLC Supplier No. HY11239210MG
GSK-3 Inhibitor XIII is a potent and ATP-competitive GSK-3 inhibitor with a Ki of 24 nM.
  • Potent and ATP-competitive GSK-3 inhibitor
  • Ki of 24 nM
  • White to off-white solid appearance
  • High purity of 98.79%
  • Soluble in DMSO (100 mg/mL)