Learn More
TARGETMOL CHEMICALS INC GSK-3 INHIBITOR XIII 5MG
SDP
Catalog No. 502576277
Encompass_Preferred
Also available in 1mg 2mg 10mg 25mg 50mg 100mg and bulk. Please contact Fisher for quotes. GSK-3 Inhibitor XIII an ATP-competitive GSK-3 inhibitor (Ki: 24 nM) can be used to study diabetes and obesity. purity: 98%
Catalog No. 50-257-6277 Supplier TARGETMOL CHEMICALS INC Supplier No. T785785MG
Add to Cart
Add to Cart
Also available in 1mg 2mg 10mg 25mg 50mg 100mg and bulk. Please contact Fisher for quotes. GSK-3 Inhibitor XIII an ATP-competitive GSK-3 inhibitor (Ki: 24 nM) can be used to study diabetes and obesity. purity: 98%