Learn More
Enzo Life Sciences GSK-3alpha/beta monoclonal antibody (1H8) (200 µg)
SDP
Supplier:  Enzo Life Sciences ADIKAMST002F
Encompass_Preferred
Clone: 1H8. Host: Mouse. Isotype: IgG2b. Immunogen: Recombinant Xenopus GSK-3β. UniProt: Q91757. Source: Purified from mouse ascites. Species Reactivity: Human, Mouse, Rat, Sheep, Rabbit, Porcine, Monkey, Hamster, Dog, Bovine. Applications: ICC, IHC, WB. Recommended dilutions: Western Blot (1:1,000, colorimetric). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-666-483