Learn More
Selleck Chemical LLC GSK-3008348 HCl-E1558-5MG
SDP
Catalog No. 503137938
Encompass_Preferred
GSK-3008348 HCl is a small molecule antagonist of integrin v 6 with an affinity (pIC50) value of 8 1 for v 6 in cell Adhesion assays
Catalog No. 50-313-7938 Supplier Selleck Chemical LLC Supplier No. E15585MG
GSK-3008348 HCl is a small molecule antagonist of integrin v 6 with an affinity (pIC50) value of 8 1 for v 6 in cell Adhesion assays