Learn More
Medchemexpress LLC GSK 3008348 hydrochloride | 1629249-40-6 | 98.0% | 10 MG
SDP
Supplier:  Medchemexpress LLC HY19767A10MG
Encompass_Preferred
GSK 3008348 hydrochloride is a small-molecule antagonist of the αvβ6 integrin used as a research tool to study integrin-mediated TGF-β activation, pulmonary fibrosis, and integrin-dependent viral entry. It exhibits low-nanomolar activity in cell assays and is supplied as a hydrochloride salt for laboratory research.
  • High potency against αvβ6 integrin with low-nanomolar IC50.
  • Selective inhibition of αv-containing integrins in cell assays.
  • Suitable for pulmonary fibrosis and oncology research applications.
  • Supplied as a hydrochloride salt with recommended storage conditions.
  • Provided in research-scale pack sizes for laboratory use.
Catalog No. 50-002-94919