Learn More
Medchemexpress LLC GSK 3008348 hydrochloride | 1629249-40-6 | 99.86% | 524.109 g/mol | 5 MG
SDP
Catalog No. 5000400462
Encompass_Preferred
GSK 3008348 hydrochloride is a small molecule antagonist of integrin αvβ6 with IC50 values for αvβ6, αvβ1, αvβ3, αvβ5, and αvβ8 of 1.50, 2.83, 12.53, 4.00, and 2.26 nM, respectively. It targets the αvβ6 integrin, inhibits TGF-β activation, and thereby alleviates the fibrotic process. It also prevents the binding of the foot-and-mouth disease virus (FMDV) to the αvβ6 integrin receptor on the surface of host cells, inhibiting viral entry. It is suitable for research on pulmonary fibrosis and various types of cancer.
  • Small molecule antagonist of integrin αvβ6.
  • Targets αvβ6 integrin.
  • Inhibits TGF-β activation, which helps alleviate fibrotic processes.
  • Prevents foot-and-mouth disease virus (FMDV) binding to the αvβ6 integrin receptor, thereby inhibiting viral entry.
  • Can be utilized in research concerning pulmonary fibrosis and various forms of cancer.
Catalog No. 50-004-00462 Supplier Medchemexpress LLC Supplier No. HY19767A5MG
GSK 3008348 hydrochloride is a small molecule antagonist of integrin αvβ6 with IC50 values for αvβ6, αvβ1, αvβ3, αvβ5, and αvβ8 of 1.50, 2.83, 12.53, 4.00, and 2.26 nM, respectively. It targets the αvβ6 integrin, inhibits TGF-β activation, and thereby alleviates the fibrotic process. It also prevents the binding of the foot-and-mouth disease virus (FMDV) to the αvβ6 integrin receptor on the surface of host cells, inhibiting viral entry. It is suitable for research on pulmonary fibrosis and various types of cancer.
  • Small molecule antagonist of integrin αvβ6.
  • Targets αvβ6 integrin.
  • Inhibits TGF-β activation, which helps alleviate fibrotic processes.
  • Prevents foot-and-mouth disease virus (FMDV) binding to the αvβ6 integrin receptor, thereby inhibiting viral entry.
  • Can be utilized in research concerning pulmonary fibrosis and various forms of cancer.