Learn More
Medchemexpress LLC Gsk 3008348 hydrochloride | 1629249-40-6 | 99.9% | 25 MG
SDP
Catalog No. 5000403401
Encompass_Preferred
GSK 3008348 hydrochloride is a small molecule antagonist of integrin αvβ6. It inhibits TGF-β activation, alleviating the fibrotic process, and prevents foot-and-mouth disease virus entry by targeting the αvβ6 integrin receptor. This compound is used in research for pulmonary fibrosis and various types of cancer.
  • Antagonizes integrin αvβ6
  • Inhibits TGF-β activation
  • Alleviates the fibrotic process
  • Prevents foot-and-mouth disease virus binding to the αvβ6 integrin receptor
  • Supports research on pulmonary fibrosis
  • Supports research on various types of cancer
Catalog No. 50-004-03401 Supplier Medchemexpress LLC Supplier No. 25MGHY19767A
GSK 3008348 hydrochloride is a small molecule antagonist of integrin αvβ6. It inhibits TGF-β activation, alleviating the fibrotic process, and prevents foot-and-mouth disease virus entry by targeting the αvβ6 integrin receptor. This compound is used in research for pulmonary fibrosis and various types of cancer.
  • Antagonizes integrin αvβ6
  • Inhibits TGF-β activation
  • Alleviates the fibrotic process
  • Prevents foot-and-mouth disease virus binding to the αvβ6 integrin receptor
  • Supports research on pulmonary fibrosis
  • Supports research on various types of cancer