Learn More
Medchemexpress LLC GSK-3484862 10MG
SDP
Catalog No. NC1862782
Encompass_Preferred
MEDCHEMEXPRESS LLC|GSK-3484862 is a non-covalent inhibitor for Dnmt1. GSK-3484862 induces DNA hypomethylation to against cancer(1)|10mg|HY-135146|Purity: 99.41%|2170136-65-7|
Catalog No. NC1862782 Supplier Medchemexpress LLC Supplier No. HY13514610MG
MEDCHEMEXPRESS LLC|GSK-3484862 is a non-covalent inhibitor for Dnmt1. GSK-3484862 induces DNA hypomethylation to against cancer(1)|10mg|HY-135146|Purity: 99.41%|2170136-65-7|