Learn More
Medchemexpress LLC HY-135146 50mg , GSK-3484862 CAS:2170136-65-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13514650MG
Encompass_Preferred
HY-135146 50mg GSK-3484862 CAS: 2170136-65-7 GSK-3484862 is a non-covalent inhibitor for DNA methyltransferase (Dnmt1). GSK-3484862 induces DNA hypomethylation to against cancer. GSK-3484862 mediates dramatic demethylation in murine embryonic stem cells with minimal non-specific toxicity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9497