Learn More
Medchemexpress LLC HY-119171 1mg Medchemexpress, GSK 366 CAS:1953157-39-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1191711MG
Encompass_Preferred
Medchemexpress, HY-119171 1mg GSK 366 CAS:1953157-39-5 GSK 366 is a potent kynurenine-3-monooxygenase (KMO) inhibitor with IC50s of 2.3 nM and 0.7 nM for human KMO and P. fluorescens-KMO (Pf-KMO), respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9516